Savant™ SpeedVac™ SPD2030A Integrated Vacuum Concentrator System
Lieferant: THERMO SAVANT
SPD2030A-230
THSASPD2030A-230EA 28300 EUR
THSASPD2030A-230
Savant™ SpeedVac™ SPD2030A Integrated Vacuum Concentrator System
Konzentratoren
Thermo Scientific™ Savant™ SpeedVac™ SPD2030 vacuum concentrators are fully integrated systems that combine a concentrator, refrigerated cold trap, diaphragm pump and a vacuum gauge into a single, compact unit.
  • Proven reliability: Over 50 years of experience with an oil-free PTFE diaphragm vacuum pump for virtually maintenance-free operation
  • Speed and efficiency: Rapid evapouration and solvent removal with a radiant cover and vacuum controls to prevent sample bumping
  • Ease of use: Intuitive design with a bright display, three preset and three user-defined programs, and an oil-free vacuum source. USB port for data transfer
  • Safety assurance: Soft-close glass lids lock securely for the safety of samples and technicians
Savant SpeedVac SPD2030 system accommodates higher capacity (i.e., 200×1.5 ml tubes and other formats). A variety of rotors are available for both systems to support sample formats from micro-centrifuge tubes, vials, and microplates to large-volume flasks. Both systems fit in a standard laboratory fume hood.
Bestellinformation: SPD2030A-230 includes acid-resistant SpeedVac™ vacuum concentrator, glass condensation flask, flask cover for GCF400. Rotor to be ordered separately.