Secure cabinets for pesticides product
Lieferant: TRIONYX
AL302
TRIOAL302EA 663 EUR
TRIOAL302
Secure cabinets for pesticides product
Möbel Lagerschränke
The cabinets is recommended for storage of herbicides, fungicides, insecticides and other phytosanitary products. Also recommended for farmers, ranchers, growers, landscapers, growers, agricultural cooperatives, green spaces, community etc. It allows a safety storage of phytosanitary products.
  • Hight and low vents
  • Double door with two key locking points
Standards: Large warning labels in compliance with ISO 3864 norm and 92/58/CEE european directive.