Sample loops for high pressure valves, stainless steel, Upchurch Scientific®
Lieferant: UPCHURCH SCIENTIFIC
1876
UPCH1876EA 532 EUR
UPCH1876
Sample loops for high pressure valves, stainless steel, Upchurch Scientific®
Chromatographie-Zubehör Chromatographie-Probenschläuche
These high quality stainless steel sample loops have burr-free, square-cut ends to ensure a flush connection to valve ports.
  • Size designations of loops are nominal
  • Supplied with fittings that are not swaged onto the tube
The loop has to be completely bottomed in the injector port before the ferrule is swaged onto the tube.