Ammoniumperoxodisulfat (APS)

Lieferant: Uptima
Gefahr

Synonyme: Peroxidisulphuric acid diammonium salt, Ammonium persulphate

UP306098
UPTIUP306098EA 0 EUR
UPTIUP306098
Ammoniumperoxodisulfat (APS)
Ammonium peroxodisulphate (APS)
Formel: (NH₄)₂S₂O₈
Molecular Weight: 228,2 g/mol
Schmelzpunkt: 120 °C
Dichte: 1,98 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003390
CAS-Nummer: 7727-54-0
EINECS: 231-786-5
UN: 1444
ADR: 5.1,III
Merck Index: 13,00545
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82a373f21d34e63dc94c0598f3f33ba6bbfdb2b&vid=2b6d196dbf07ce4dd528ac7b4f8456678accd3ba&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR