N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimid Hydrochlorid
Lieferant: Uptima
|
Achtung
|
Synonyme:
N'-(Ethylcarbonimidoyl)-N,N-dimethylpropane-1,3-diamine monohydrochloride, N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide monohydrochloride, 1-Ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride, EDC hydrochloride, EDC, EDAC HCl, EDAC hydrochloride, 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, WSC HCl, EDC HCl
UP52005B
UPTIUP52005BEA
0
EUR
UPTIUP52005B
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimid Hydrochlorid
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride
Formel:
C₈H₁₇N₃·HCl Molecular Weight: 191,7 g/mol Schmelzpunkt: 112…113 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00012503 CAS-Nummer: 25952-53-8 EINECS: 247-361-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...