Bosshead (sleeve)
Lieferant: USBECK Laborgeräte
2240
USBE2240EA 6.6 EUR
USBE2240
Bosshead (sleeve)
Klemmen Stativmuffen
Bosshead made of powder-coated zinc die-cast, with nickel plated brass screws.
  • For rods up to 16 mm Ø
  • Location hole 10 mm Ø
Weight: 80 g