Gel documentation systems, ChromaDoc-It™
Lieferant: UVP ULTRA VIOLET PRODUCTS
97-0109-02
UVPA97-0109-02EA 7550 EUR
UVPA97-0109-02
Gel documentation systems, ChromaDoc-It™
Photodokumentations-Systeme
The ChromaDoc-It™ system connects to a computer for acquisition and documentation of thin layer chromatography (TLC) images. The high resolution Digi camera is controlled by UVP's exclusive capture software.
  • DigiCam 130 digital colour camera with 17,9 megapixel resolution
  • Low profile Chromato-Vue UV viewing cabinet (15 W) includes overhead UV (254/365 nm)
  • Capture software controls camera functions, image capture and image enhancement; allows live preview of images
Application: Thin Layer Chromatography (TLC) gel imaging with overhead UV lamps.
Bitte beachten UVP software requires Microsoft Windows 8, Windows 7 or Vista (32-bit or 64-bit).