Nitrogen analyser
Lieferant: VELP SCIENTIFIC
F30800080
VELPF30800080EA 0 EUR
VELPF30800080
Nitrogen analyser
Analysatoren Protein-Analysatoren
Powerful protein/nitrogen analyser able to perform precise analysis and determination in a flash for high productivity and unique benefits.
  • Controllable from PC with DUMAsoft™ software for detailed reporting
  • Automated analysis of preloaded samples for over 100 positions
  • User-selectable methods with storable data
Standards: Conforms to Good Laboratory Practice standard.