Tygon S3™ B-44-3 Tubing
Lieferant: Saint Gobain Life Sciences
AAB00046
VERNAAB00046EA 1040 EUR
VERNAAB00046
Tygon S3™ B-44-3 Tubing
Schläuche Transferschlauch
Tygon S3™ B-44-3 tubing for beverage transfer from Saint-Gobain is phthalate-free and specifically formulated to ensure it does not affect the taste nor odour of liquids moving through it, including soft drinks and juices.
  • Clear as glass for easy visual monitoring of flow
  • Lightweight and flexible for easy, quick installation
  • Broad chemical resistance
  • Non-wetting properties allow easy cleaning and complete drainage
  • Contains no BPA or phthalates
  • Autoclavable
It is non-wetting, which allows it to completely drain and flush clean with ease, and its glass-like clarity makes it easy to monitor and adjust the beverage flow during processing. Light in weight and easy to handle, Tygon S3 B-44-3 tubing can be put into service quickly. It readily bends to accommodate abrupt corners and obstructions, requiring a minimium of couplings and fittings. It is ideal for applications such as food and beverage transfer and dispensing.
Hardness: 63 (shore A)