Formaldehyd 24% in wässriger Lösung
Lieferant: KLINIPATH
Synonyme:
Methanal, Formalin, Formol, Methylaldehyd
VWRK4094-9020EA
191
EUR
VWRK4094-9020
Formaldehyd 24% in wässriger Lösung
Formaldehyd
Formel:
CH₂O Molecular Weight: 30,03 g/mol |
MDL:
MFCD00003274 CAS-Nummer: 50-00-0 UN: 3334 ADR: 9, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...