Formaldehyd 4%, gepuffert

Lieferant: KLINIPATH

Synonyme: Methanal, Formalin, Formol, Methylaldehyd

4187F
VWRK4187FEA 154 EUR
VWRK4187F
Formaldehyd 4%, gepuffert
Formaldehyd

Formaldehyde solution 4% (pH = 7).


  • Pre-filled solutions


For use as a general purpose histological fixative.


Verpackung: In ready-to-use screw cap PTFE or glass containers.

Formel: CH₂O
Molecular Weight: 30,03 g/mol
MDL: MFCD00003274
CAS-Nummer: 50-00-0
IVDLogo

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR