Control Standard Endotoxin (CSE)
Lieferant: FUJIFILM WAKO CHEMICALS
CSE4037-5006
WAKFCSE4037-5006EA
126
EUR
WAKFCSE4037-5006
Control Standard Endotoxin (CSE)
Mikrobiologische Testsysteme
Endotoxin-Tests
The Control Standard Endotoxin (CSE) is a lyophilised product composed of a 500 ng vial of endotoxin, purified from E. coli UKT-B strain that serves as an accurate and dependable standard for endotoxin testing.
- Endotoxin derived from E. coli UKT-B
- Can be used to prepare controls and standard curves
- Potency of CSE supplied to match each lot of LAL
- Reconstituted CSE can be stored at 2 to 10 °C for 1 month