WeLoc® Gripper PA 150 Bag Closures
Lieferant: WELOC
S1915004
WELOS1915004EA 499 EUR
WELOS1915004
WeLoc® Gripper PA 150 Bag Closures
Klemmen Klemmen für den allgemeinen Gebrauch
The WeLoc Gripper PA is the most powerful WeLoc closure. It is manufactured from DuPont's nylon, reinforced with glass fibre which further enhances its strength and durability.
  • Material: Polyamide reinforced with glass fibre
  • Temperature ranges: –20 to +120 ⁰C
  • Sterilisation methods: Autoclave, EtO, Gamma
  • Weight: 130 g
The gripper is designed to cope with the toughest environments and is therefore excellent for many industrial applications, where a stronger seal is often required.
Verpackung: Pieces/carton: 70, Pieces/pallet: 2800 (Weight/carton: 9,6 kg, Weight/pallet: 409 kg).