Wright-Giemsa stain modifiziert, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
Wright-Giemsa Solution
WG32-1L
WG32-1LEA
254
EUR
WG32-1L
Wright-Giemsa stain modifiziert, Sigma-Aldrich®
Wright-Giemsa stain (solutions & kits)
When blood films are stained using Wright-Giemsa Stain, the white blood cell nucleus and cytoplasm take on the characteristic blue or pink coloration. The combination of purified eosin and thiazine dyes in the product eliminates inconsistent staining and yields reproducible chromogenic responses.