Glass fibre tape, HGF61, Whatman™
Lieferant: Whatman products (Cytiva)
1830-6236
WHAT1830-6236EA 35.9 EUR
WHAT1830-6236
Glass fibre tape, HGF61, Whatman™
Filter Glasfaserfilter
HGF61 glass fibre tape provides strength, efficiency, reliability and consistency of data quality in continuous air monitoring.
  • Meets regulatory requirements for continuous air particulate sampling globally
  • Retains 99,99% of air particulate at 0,3 µm - each lot is quality tested
  • Suitable for PM1, PM2.5, PM10 and TSP sampling and for beta-attenuation method
  • Provides excellent consitency of data quality in air sampling
  • Effective in both humid and dry environments
  • Compatable with most continuous air sampling devices
HGF61 glass fiber tape with organic binder has excellent mechanical strength without any fiber shedding.