Turbine spinner for use with Bruker® Spectrometers, SP Wilmad-LabGlass
Lieferant: SP Industries
B-PEEK-5
WILMB-PEEK-5EA 650 EUR
WILMB-PEEK-5
Turbine spinner for use with Bruker® Spectrometers, SP Wilmad-LabGlass
Rührer Rührwerkzeuge
Manufactured to precision standards that exceed the spectrometer manufacturers requirements.
  • Variable temperature operation of -150 to +200 °C
  • Weight compatible with room temperature spinners
  • Susceptibility matched with Bruker® originals
Can be mixed with other turbines during sample changer operation.