Bulb pipettes, Class A, SP Wilmad-LabGlass
Lieferant: SP Industries
LG-9350-136
WILMLG-9350-136EA 137 EUR
WILMLG-9350-136
Bulb pipettes, Class A, SP Wilmad-LabGlass
Pipetten Vollpipetten
Glass.
  • Calibrated to deliver at 20 °C
  • Fabricated to meet the requirements of accuracy, appearance, glass quality and calibration line
  • Colour coded
Standards: ASTM E-969.