Extractor, for light solvents
Lieferant: WINZER LABORGLASTECHN
21-550-81
WINZ21-550-81EA 0 EUR
WINZ21-550-81
Extractor, for light solvents
Extraktoren Flüssig-Flüssig-Extraktoren
Made of borosilicate glass. Extractor specially designed to extract liquids for light solvents.
  • Very good chemical resistance
  • Very good temperature resistance
This unit is designed with valve stopcock with 0,3 mm PTFE spindle.