Standard ground joints, sockets
Lieferant: WITEG LABORTECHNIK
0.102.060
WITG0.102.060EA 29.7 EUR
WITG0.102.060
Standard ground joints, sockets
Adapter Übergangsstücke mit Kegelschliffen
Made of borosilicate glass.
  • Very good temperature resistance
  • Very good chemical resistance
Standards: DIN 12 249.