Woulff bottles
Lieferant: WITEG LABORTECHNIK
5.802.001
WITG5.802.001EA 173 EUR
WITG5.802.001
Woulff bottles
Flaschen Woulffsche Flaschen
Made of borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion gives relatively high resistance to temperature changes
Two side necks with GL 18 thread, center neck with GL 25 thread. With bottom tubulature with screw cap and olive for tubing connection.