Wright Stain Set Set for Hema-Tek® I, 1000 and 2000, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

WSHT-1SET
WSHT-1SETEA 111 EUR
WSHT-1SET
Wright Stain Set Set for Hema-Tek® I, 1000 and 2000, Sigma-Aldrich®
Wright Stain Set

Specific packaging intended for use on the Hema-Tek and Hema-Tek 1000 hematology stainers.

Lagertemperatur: Raumtemperatur
ADR: 3,II
IVDLogo

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR