ORP electrode, TA-SensoLyt PtA
Lieferant: WTW
103711
WTWA103711EA 437 EUR
WTWA103711
ORP electrode, TA-SensoLyt PtA
Elektroden Redox-Elektroden
ORP combination electrode (platinum) for installation in depth armature series TA 197-pH, polymer electrolyte, pin hole junction.