Carbonate buffer concentrate
Lieferant: YSI LIFE SCIENCE
1579
YSIL1579EA
0
EUR
YSIL1579
Carbonate buffer concentrate
Puffer
Biologische Puffer
Carbonate buffer concentrate for use with methanol membranes and ethanol membranes. Reconstitute to make 1 litre.
- Equipment used with: 2900, 7100, 2700, 2950
Bitte beachten Store at 4 to 25 °C