Kaliumhexacyanoferrat(II)
Lieferant: YSI LIFE SCIENCE
|
|
Synonyme:
Tetrakaliumhexacyanoferrat, Ferrocyankalium
2363
YSIL2363EA
0
EUR
YSIL2363
Kaliumhexacyanoferrat(II)
Kaliumhexacyanoferrat(II)
Formel:
C₆FeK₄N₆ Molecular Weight: 368,35 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00167022 CAS-Nummer: 13943-58-3 EINECS: 237-722-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...