Biochemistry analyser
Lieferant: YSI LIFE SCIENCE
2900D
YSIL2900DEA 0 EUR
YSIL2900D
Biochemistry analyser
Analysatoren Elementar-Analysatoren
Biochemistry analyser features an intuitive graphical user interface, a USB port for data retrieval to measure samples from sample holders including 96 well plates and microcentrifuge tubes.
  • Analyse specific results in a minute or less
  • Proprietary immobilised enzyme electrodes
  • Unique fluidics resist clogging and automated sample handling
Measures the chemistries in application areas such as glucose, sucrose, lactate, galactose, glutamine, lactose, glutamate, choline, xylose, glycerol, ethanol, methanol, hydrogen peroxide.
Standards: ETL, RoHS