Wipes for general applications, Polytex® light
Lieferant: Zellstoff-Vertriebs-GmbH & Co. KG
10160-00
ZVGA10160-00EA 144 EUR
ZVGA10160-00
Wipes for general applications, Polytex® light
Wischtücher Reinigungstücher, wiederverwendbar Tücher für allgemeine Anwendungen
These solvent resistant wipes made of 100% PP are suitable for wet wiping in a variety of applications.
  • Durable and lint-free
  • Even remove stubborn dirt
  • Suitable for food contact
Ideal for use in garages, paint shops, catering areas, kitchens and food processing areas.