VWR®, Safety Storage Cabinets for Flammables
Lieferant: VWR
VWR®
VWRU89522-626 VWRU89522-632 VWRU89522-622
VWR®, Safety Storage Cabinets for Flammables
Möbel Lagerschränke
VWR® safety cabinets for flammables are designed to protect both laboratory personnel and the cabinet contents.
  • Fully welded, 18-gauge double-wall construction for fire protection
  • Upper and lower diametrically opposed side vents
  • All models fire tested and approved by FM Global
Fully welded, 18-gauge galvanised steel ensures fire protection and strength with reduced gaps. Equipped with upper and lower diametrically opposed side vents, a ground connector on lower right side of the cabinet, double-wall construction with 38 mm insulating air space, and an environmentally friendly, electrostatically applied powder coat finish.
Standards: All door styles meet OSHA 1910.106, NFPA 30 and NFPA 1. Self-close door styles also meet IFC requirements and feature a fusible link that melts at 74 °C for automatic closure in the event of a fire.