Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')kupfer(II), Sigma-Aldrich®

Lieferant: Merck

SIAL345083-1GEA 49 EUR
SIAL345083-1G
Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')kupfer(II), Sigma-Aldrich®
Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')kupfer(II)
Formel: C₂₂H₃₈CuO₄
Molecular Weight: 430,08 g/mol
MDL: MFCD00058920
CAS-Nummer: 14040-05-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR