Tricyclohexylphosphin 1 mol/l in Toluol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

SIAL592285-100MLEA 418 EUR
SIAL592285-100ML
Tricyclohexylphosphin 1 mol/l in Toluol, Sigma-Aldrich®
Tricyclohexylphosphine
Formel: C₁₈H₃₃P
Molecular Weight: 280,43 g/mol
MDL: MFCD00003853
CAS-Nummer: 2622-14-2
EINECS: 220-069-2
UN: 1294
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR