Fmoc-L-phenylalanin

Lieferant: Molekula

MOLEM37869179EA 0 EUR
MOLEM37869179
Fmoc-L-phenylalanin
Fmoc-L-phenylalanin
Formel: C₂₄H₂₁NO₄
Molecular Weight: 387,4 g/mol
MDL: MFCD00037128
CAS-Nummer: 35661-40-6
EINECS: 252-661-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR