Tricain-Methansulfonat

Lieferant: Serva

SERA12396.02EA 63.2 EUR
SERA12396.02 SERA12396.03
Tricain-Methansulfonat
Tricain-Methansulfonat
Formel: C₁₀H₁₅NO₅S
Molecular Weight: 261,29 g/mol
MDL: MFCD00013176
CAS-Nummer: 886-86-2
EINECS: 212-956-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR