L-Glutaminsäure-5-methylester, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL858269-25GEA 106 EUR
SIAL858269-25G
L-Glutaminsäure-5-methylester, Sigma-Aldrich®
L-Glutaminsäure-5-methylester
Formel: C₆H₁₁NO₄
Molecular Weight: 161,2 g/mol
MDL: MFCD00002632
CAS-Nummer: 1499-55-4
EINECS: 216-110-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR