N-Fmoc-glycin

Lieferant: Molekula

MOLE18662074-25GEA 68.7 EUR
MOLE18662074-25G
N-Fmoc-glycin
N-Fmoc-glycin
Formel: C₁₇H₁₅NO₄
Molecular Weight: 297,3 g/mol
MDL: MFCD00037140
CAS-Nummer: 29022-11-5
EINECS: 249-373-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR