N-Fmoc-glycin

Lieferant: NOVABIOCHEM

Synonyme: 2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)acetic acid, Fmoc-Gly-OH

8.52001.0100 8.52001.0025
04-12-1001-100EA 98.9 EUR
04-12-1001-100 04-12-1001-25
N-Fmoc-glycin
N-Fmoc-glycin
Formel: C₁₇H₁₅NO₄
Molecular Weight: 297,3 g/mol
MDL: MFCD00037140
CAS-Nummer: 29022-11-5
EINECS: 249-373-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR