1-Amino-2-methyl-9,10-anthrachinon
Lieferant: EHRENSTORFER GMBH
EHERC12972111EA
85
EUR
EHERC12972111
1-Amino-2-methyl-9,10-anthrachinon
1-Amino-2-methyl-9,10-anthrachinon
Formel:
C₁₅H₁₁NO₂ |
MDL:
MFCD00001220 CAS-Nummer: 82-28-0 EINECS: 201-408-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...