2-(Aminomethyl)-5-brompyridin

Lieferant: BEPHARM LTD.

BEPHB160375EA 0 EUR
BEPHB160375
2-(Aminomethyl)-5-brompyridin
2-(Aminomethyl)-5-bromopyridine
Formel: C₆H₇BrN₂
Molecular Weight: 187,0400 g/mol
MDL: MFCD06213858
CAS-Nummer: 173999-23-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR