Dibenzylphosphat

Lieferant: CHEMPUR

CHMPFL13507.POREA 0 EUR
CHMPFL13507.POR
Dibenzylphosphat
Dibenzylphosphat
Formel: C₁₄H₁₅O₄P
Molecular Weight: 278,24 g/mol
MDL: MFCD00004775
CAS-Nummer: 1623-08-1
EINECS: 216-602-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR