Divinylsulfon
Lieferant: FLUOROCHEM LTD (CHEMPUR)
FLUO008036-500GEA
0
EUR
FLUO008036-500G
FLUO008036-25G
FLUO008036-100G
Divinylsulfon
Divinylsulfon
Formel:
C₄H₆O₂S Molecular Weight: 118,16 g/mol |
MDL:
MFCD00008623 CAS-Nummer: 77-77-0 EINECS: 201-057-6 UN: 2927 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...