Divinylsulfon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIALV3700-100GEA 457 EUR
SIALV3700-100G SIALV3700-25G SIALV3700-5G
Divinylsulfon, Sigma-Aldrich®
Divinylsulfon
Formel: C₄H₆O₂S
Molecular Weight: 118,16 g/mol
MDL: MFCD00008623
CAS-Nummer: 77-77-0
EINECS: 201-057-6
UN: 2927
ADR: 6.1,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR