2-Pyridinsulfonsäure, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ200
SIAL661759-10GEA
302
EUR
SIAL661759-10G
2-Pyridinsulfonsäure, Sigma-Aldrich®
2-Pyridinesulphonic acid
Formel:
C₅H₅NO₃S Molecular Weight: 159,1631 g/mol |
MDL:
MFCD00234080 CAS-Nummer: 15103-48-7 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...