Digitonin, wasserlöslich

Lieferant: Serva

SERA19551.01EA 210 EUR
SERA19551.01 SERA19551.02
Digitonin, wasserlöslich
Digitonin
Formel: C₅₆H₉₂O₂₉
Molecular Weight: 1229,312 g/mol
MDL: MFCD00077729
CAS-Nummer: 11024-24-1
EINECS: 234-255-6
UN: 2811
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR