3-Decanol ≥97%

Lieferant: Thermo Scientific

B24141.06 B24141.14
B24141.06EA 82.2 EUR
B24141.06 B24141.14
3-Decanol ≥97%
3-Decanol

Purity: 97% CAS No.: 1565-81-7 Molecular Formula: C10H22O Formula Weight: 158.29 Flash Point: 99°(210°F) Refractive Index: 1.4350 Boiling Point: 213° Beilstein Reference: 1719658 MDL No.: MFCD00021956

Formel: C₁₀H₂₂O
Molecular Weight: 158,29 g/mol
Siedepunkt: 213 °C
Dichte: 0,830
Flammpunkt: 99 °C (210 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00021956
CAS-Nummer: 1565-81-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR