Glycin (H-Gly-OH) ≥99,5% für die Elektrophorese
Lieferant: Thermo Scientific
Synonyme: Aminoacetic acid, Glycocoll, H-Gly-OH
J64365.30 J64365.A1
J64365.30EA 41 EUR
J64365.30 J64365.A1
Glycin (H-Gly-OH) ≥99,5% für die Elektrophorese
Glycine
Purity: 99.5+% CAS No.: 56-40-6 Molecular Formula: C2H5NO2 Formula Weight: 75.07 Physical Form: Powder Density: 1.595 Appearance: White Odour: Odourless pH: 5,97 Melting Point: ca 245° dec. Solubility: Freely soluble in water Beilstein Reference: 635782 Merck Reference: 14,4491 MDL No.: MFCD00008131
Formel: NH₂CH₂COOH
Molecular Weight: 75,07 g/mol
Siedepunkt: 181 °C (1013 hPa)
Schmelzpunkt: ∼ 245 °C (Zers.)
Dichte: 1,595
Lagertemperatur: Raumtemperatur
MDL: MFCD00008131
CAS-Nummer: 56-40-6
EINECS: 200-272-2
Merck Index: 14,4491
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM