D(+)-Glucose Monohydrat, Calbiochem®
Lieferant: Merck Millipore Calbiochem
Synonyme:
Dextrose Monohydrat
, Traubenzucker Monohydrat
4074-2EA
94.4
EUR
4074-2
D(+)-Glucose Monohydrat, Calbiochem®
D(+)-Glucose Monohydrat
Formel:
C₆H₁₂O₆·H₂O Molecular Weight: 198,17 g/mol Schmelzpunkt: 83 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149450 CAS-Nummer: 14431-43-7 EINECS: 200-075-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...