Polyvinylpyrrolidon, M.W. ∼ 40,000 (K30)
Lieferant: Thermo Scientific
Synonyme: PolyvidonPoly[1-(2-oxo-1-pyrrolidinyl)ethylen]PVP
J62417.36EA 138 EUR
J62417.36 J62417.22 J62417.A1
Polyvinylpyrrolidon, M.W. ∼ 40,000 (K30)
Polyvinylpyrrolidon
A cryoprotectant, also useful in plant cell culture research
Formel: (C₆H₉NO)n
Lagertemperatur: Raumtemperatur
MDL: MFCD01076626
CAS-Nummer: 9003-39-8
Merck Index: 14,7697