Rhodamin 6G
Lieferant: EHRENSTORFER GMBH
Synonyme:
9-[2-(Ethoxycarbonyl)phenyl]-3,6-bis(ethylamino)-2,7-dimethylxanthyliumchlorid
EHERC10424950EA
82
EUR
EHERC10424950
Rhodamin 6G
Rhodamin 6G
Formel:
C₂₈H₃₁ClN₂O₃ Molecular Weight: 479,02 g/mol Siedepunkt: ∼200 °C (1013 hPa) |
MDL:
MFCD00012665 CAS-Nummer: 989-38-8 EINECS: 213-584-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...