5,8,11,14-Eicosatetrainsäure

Lieferant: Thermo Scientific

J61624.MCEA 587 EUR
J61624.MC J61624.FX
5,8,11,14-Eicosatetrainsäure
5,8,11,14-Eicosatetraynoic acid

A cyclooxygenase, lipoxygenase and cytochrome P450 inhibitor

Formel: C₂₀H₂₄O₂
Molecular Weight: 296,41 g/mol
Schmelzpunkt: 81…82 °C
MDL: MFCD00036967
CAS-Nummer: 1191-85-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR