Glass Microfibre Filters, Grade MGT, Munktell
Lieferant: Ahlstrom-Munksjö
BINZ440012EA 15.3 EUR
BINZ440012
Glass Microfibre Filters, Grade MGT, Munktell
Fine fibers ensure high efficiency filtration of submicron particles while the fiber matrix creates both depth filtration and good air permeability.
  • Compatible with a large number of inorganic and organic solutions
  • Physically and biologically inert
  • Efficiency of 99,95% (0,12 – 0,3 μm)