Diethylphthalat-[Ring-D4]

Lieferant: EHRENSTORFER GMBH

EHERC16172010EA 119 EUR
EHERC16172010
Diethylphthalat-[Ring-D4]
Diethylphthalat-[Ring-D4]
Formel: C₁₂H₁₀D₄O₄
MDL: MFCD00144117
CAS-Nummer: 93952-12-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR