Diethylphthalat-[Ring-D4] (98% D), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIAL492221-250MGEA 644 EUR
SIAL492221-250MG
Diethylphthalat-[Ring-D4] (98% D), Sigma-Aldrich®
Diethylphthalat-[Ring-D4]
Formel: C₁₂H₁₀D₄O₄
MDL: MFCD00144117
CAS-Nummer: 93952-12-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR